Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Anhydride, 99%

Catalog No. AC158760500
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum

Succinic anhydride, 99%, C4H4O3, CAS Number-108-30-5, 2,5-diketotetrahydrofuran, 2,5-furandione, dihydro, butanedioic anhydride, dihydro-2,5-furandione, dihydrofuran-2,5-dione, rikacid sa, succinic acid anhydride, succinic anhydride, succinyl anhydride, succinyl oxide, 1kg, 17,407, 261.0 deg.C | CAS: 108-30-5 | C4H4O3 | 100.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-30-5
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00005525
InChI Key RINCXYDBBGOEEQ-UHFFFAOYSA-N
Synonym succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
PubChem CID 7922
ChEBI CHEBI:36595
IUPAC Name oxolane-2,5-dione
SMILES O=C1CCC(=O)O1

Specifications

Melting Point 116.0°C to 120.0°C
Boiling Point 261.0°C
Quantity 50 g
Packaging Plastic bottle
Assay Percent Range 99%
Viscosity 3 mPa.s (125°C)
Beilstein 17,407
Fieser 04,468
Merck Index 15,9
Solubility Information Solubility in water: 62g/L (20°C). Other solubilities: soluble in chloroform,carbon tetrachloride and,alcohol,slightly soluble in ether,20g/L benzene (20°C)
Formula Weight 100.07
Percent Purity 99%
Chemical Name or Material Succinic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately
EINECSNumber 203-570-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.