Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18607573
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

TR117, 121-44-8, C6H15N

Spectrum™ Chemical Triethylamine, Reagent is a chemical compound that is often referred to as TEA. This volatile liquid has no color and has a pungent fish like smell combined with ammonia. Most often used as a base in organic synthesis. It is also specifically used as the active ingredient of mosquito anesthetic. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
IUPAC Name triethylamine
SMILES CCN(CC)CC

Specifications

CAS Max % 1
Packaging Steel Can
Quantity 20 L
Percent Purity 99%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.