Learn More
Solstice Triethylammonium Phosphate Solution, pH 3, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Finest quality products for life science
- The most stringent testing protocols and highest purity
- Superior manufacturing methods and product specific testing protocols designed to ensure quality to our highest standard
- Tests may include, but are not limited to, purity by gel electrophoresis, trace metal testing and application testing.
Specifications
Specifications
| CAS | 10138-93-9 |
| pH | 2.9 to 3 |
| Quantity | 100 mL |
| Packaging | Glass bottle |
| Molecular Weight (g/mol) | 199.187 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.