Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylborane, 1M soln. in THF

Catalog No. aal15109ac
Click to view available options
Quantity:
25 mL
100 mL

CAS: 97-94-9 | C6H15B | 98.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylborane is catalyst for allylation of aldehydes, decarboxylative C-C bond cleavage reactions, rhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenations, Regioselective hydroxyalkylation of unsaturated oxime ethers. Reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes and synthesis of tetramethylammonium trialkylphenylborate salts with oxidation potential.

Solubility
Reacts with water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-94-9
Molecular Formula C6H15B
Molecular Weight (g/mol) 98.00
MDL Number MFCD00009022
InChI Key LALRXNPLTWZJIJ-UHFFFAOYSA-N
Synonym triethylboron, borane, triethyl, triethylborine, boron triethyl, boron ethyl, unii-z3s980z4p3, hsdb 897, triethylboran, borethyl, triethyl borane
PubChem CID 7357
IUPAC Name triethylborane
SMILES CCB(CC)CC

Specifications

Density 0.865
Flash Point −17°C (1°F)
Linear Formula (CH3CH2)3B
Quantity 25 mL
UN Number UN2924
Beilstein 1731462
Sensitivity Air Sensitive
Solubility Information Reacts with water.
Formula Weight 98
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Triethylborane
Show More Show Less
Hazard Category H225-H302+H332-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 202-620-9
RTECSNumber ED2100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.