Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene glycol, 99%

Catalog No. p-7080996
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL

CAS: 112-27-6 | C6H14O4 | 150.17 g/mol

Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

Solubility
Miscible with water, ethanol, acetone, benzene, toluene, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil, fat, petroleum ether and most hydrocarbons.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
MDL Number MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
Synonym triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanol
PubChem CID 8172
ChEBI CHEBI:44926
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethanol
SMILES OCCOCCOCCO

Specifications

Melting Point °C to 7°C
Density 1.125 g/mL
Boiling Point 284°C to 288°C
Flash Point 165°C (329°F)
Refractive Index 1.4555
Linear Formula H(OCH2CH2)3OH
Quantity 500 mL
Beilstein 969357
Sensitivity Hygroscopic
Merck Index 14,9670
Solubility Information Miscible with water,ethanol,acetone,benzene,toluene,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil,fat,petroleum ether and most hydrocarbons.
Formula Weight 150.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethylene glycol
Show More Show Less
EINECSNumber 203-953-2
RTECSNumber YE4550000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.