Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene glycol, 99%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
112-27-6
C6H14O4
150.17
MFCD00002880
ZIBGPFATKBEMQZ-UHFFFAOYSA-N
triethylene glycol,triglycol,2,2'-ethane-1,2-diylbis oxy diethanol,trigen,triethylenglykol,triethyleneglycol,2,2'-ethylenedioxydiethanol,1,2-bis 2-hydroxyethoxy ethane,3,6-dioxaoctane-1,8-diol,2,2'-ethylenedioxy diethanol
8172
CHEBI:44926
2-[2-(2-hydroxyethoxy)ethoxy]ethanol
C(COCCOCCO)O
112-27-6
C6H14O4
150.17
MFCD00002880
ZIBGPFATKBEMQZ-UHFFFAOYSA-N
triethylene glycol,triglycol,2,2'-ethane-1,2-diylbis oxy diethanol,trigen,triethylenglykol,triethyleneglycol,2,2'-ethylenedioxydiethanol,1,2-bis 2-hydroxyethoxy ethane,3,6-dioxaoctane-1,8-diol,2,2'-ethylenedioxy diethanol
8172
CHEBI:44926
2-[2-(2-hydroxyethoxy)ethoxy]ethanol
C(COCCOCCO)O
Melting Point -7°C
Flash Point 165°C
Boiling Point 285°C
Specific Gravity 1.12
Refractive Index 1.4545 to 1.4565
Viscosity 48 mPa.s (20°C)
Infrared Spectrum Authentic
Density 1.1200g/mL
Color Yellow
Vapor Pressure <0.01mbar at 20°C
Quantity 2.5L
Packaging Glass Bottle
Assay Percent Range 99%
Linear Formula HOCH2CH2OCH2CH2OCH2CH2OH
Beilstein 01, IV, 2400
Merck Index 15, 9835
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,benzene and toluene,sparingly soluble in ether,insoluble in petroleum ether,aliphatic,hydrocarbons
Formula Weight 150.17
Physical Form Viscous Liquid
Percent Purity ≥98.5%
Chemical Name or Material Triethylene glycol
Show More Show Less