Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylene glycol dimethacrylate

Catalog No. 261548250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mL
1 L

Catalog No. 26-154-8250ML

Supplier: Sigma Aldrich 261548250ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylene glycol dimethacrylate (TEGDMA) is a hydrophilic, low viscosity, difunctional methacrylic monomer employed as a crosslinking agent.TEGDMA is a monomer typically used in dental resin materials that can cause specific stress responses in eukaryotic cells.

Specifications

CAS 109-16-0
Density 1.092 g/mL (at 25°C)
Boiling Point 170°C to 172°C (5 mmHg)
Molecular Formula C14H22O6
Refractive Index n20/D 1.461 (literature)
Linear Formula CH2=C(CH3)COO(CH2CH2O)3 COC(CH3)=CH2
Quantity 250 mL
MDL Number MFCD00008591
Molecular Weight (g/mol) 286.32
Percent Purity 95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.