Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyleneglycol Dibutylether 99%, Thermo Scientific™

Catalog No. AC339920050
Click to view available options
Quantity:
5 g

Catalog No. AC339920050

Supplier: Thermo Scientific Chemicals 339920050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C14H30O4

Specifications

Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4290 to 1.4310 (20°C, 589nm)
Linear Formula H3C(CH2)3[O(CH2)2]4CH2CH3
Quantity 5 g
Formula Weight 262.39
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Triethyleneglycol dibutylether, 99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.