Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylenetetramine-N,N,N',N″,N‴,N‴-hexaacetic Acid 98.0+%, TCI America™
SDP

Catalog No. T080625G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 869-52-3
Molecular Formula C18H30N4O12
Molecular Weight (g/mol) 494.454
MDL Number MFCD00004290
InChI Key RAEOEMDZDMCHJA-UHFFFAOYSA-N
Synonym ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acid
PubChem CID 70088
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 494.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triethylenetetramine-N,N,N′,N′′,N′′′,N′′′-hexa
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.