Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
Solubility
Soluble in Dichloromethane. Reacts with water.
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
Solubility
Soluble in Dichloromethane. Reacts with water.
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Density | 1.328 |
Odor | Chloroform-like |
Linear Formula | (CH3CH2)3OBF4 |
Quantity | 500 mL |
UN Number | UN2922 |
Beilstein | 3598090 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in Dichloromethane. Reacts with water. |
Formula Weight | 189.99 |
Concentration or Composition (by Analyte or Components) | 1.0M in dichloromethane |
Show More |
Safety and Handling
Hazard Category | H314-H336-H350 |
Hazard Statement | GHS H Statement H314-H318-H351 Causes severe skin burns and eye damage. Causes serious eye damage. Suspected of causing cancer. |
Precautionary Statement | P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 206-705-1 |
RTECSNumber | RR4584700 |
TSCA | Yes |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction