Learn More
Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether, Thermo Scientific Chemicals
Description
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of μ -amino esters from lactams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of µ -amino esters from lactams.
Solubility
Soluble in dichloromethane.
Notes
Moisture sensitive and hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 84°C to 89°C |
Assay Percent Range | 95%, Stabilized with 3-5% diethyl ether |
Linear Formula | (CH3CH2)3OBF4 |
Quantity | 25 g |
UN Number | UN2925 |
Beilstein | 3598090 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dichloromethane. |
Formula Weight | 189.99 |
Percent Purity | 95% |
Show More |
Safety and Handling
Hazard Category | H314-H335-H336 |
Hazard Statement | GHS H Statement H228-H314 |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S. |
EINECSNumber | 206-705-1 |
RTECSNumber | RR4584700 |
TSCA | Yes |
storageNote1 | Moisture sensitive |
Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.