Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylphosphine, 97%

Catalog No. p-7049839
Click to view available options
Quantity:
5 g
25 g

CAS: 554-70-1 | C6H15P | 118.16 g/mol

Triethylphosphine is used as a ligand for arylation. It is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-70-1
Molecular Formula C6H15P
Molecular Weight (g/mol) 118.16
MDL Number MFCD00009256
InChI Key RXJKFRMDXUJTEX-UHFFFAOYSA-N
Synonym triethylphosphine, phosphine, triethyl, triethylphosphorus, triethyl phosphine, pet3, et3p, unii-5w435d16pm, 3ep, triethylphophine, phosphorus triethyl
PubChem CID 27365
ChEBI CHEBI:39971
IUPAC Name triethylphosphane
SMILES CCP(CC)CC

Specifications

Density 0.81 g/mL
Boiling Point 126°C to 128°C
Assay Percent Range 97%
Quantity 5 g
UN Number UN2845
Sensitivity Air and moisture sensitive
Merck Index 14,9675
Solubility Information Insoluble in water.
Formula Weight 118.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethylphosphine
Show More Show Less
Hazard Category H225-H250-H252-H314-H332-H335
Hazard Statement GHS H Statement
H225-H250-H252-H332
Precautionary Statement P210-P222-P233-P235-P235+P410-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 209-068-8
RTECSNumber SZ3400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.