Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylsilane 98.0+%, TCI America™
SDP

Catalog No. T0662250ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
250 mL

Chemical Identifiers

CAS 617-86-7
Molecular Formula C6H15Si
Molecular Weight (g/mol) 115.271
MDL Number MFCD00009018
InChI Key QXTIBZLKQPJVII-UHFFFAOYSA-N
Synonym triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
PubChem CID 6327258
IUPAC Name triethylsilicon
SMILES CC[Si](CC)CC

Specifications

Boiling Point 108°C
Color Colorless
Physical Form Liquid
Quantity 250 mL
UN Number 1993
Formula Weight 116.28
Percent Purity ≥98.0% (GC)
Chemical Name or Material Triethylsilane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.