Learn More
Triethylsulfonium bis(trifluoromethylsulfonyl)imide, 98%
Description
Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Quantity | 50 g |
| Formula Weight | 399.39 |
| Percent Purity | 98% |
| Chemical Name or Material | Triethylsulfonium bis(trifluoromethylsulfonyl)imide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.