Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIF Monoclonal Antibody (OTI1G7)
GREENER_CHOICE

Catalog No. PIMA526477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526477 Supplier Invitrogen™ Supplier No. MA526477
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIF Monoclonal Antibody for Western Blot

Involved in innate immunity against invading pathogens. Adapter used by TLR3 and TLR4 (through TICAM2) to mediate NF-kappa-B and interferon-regulatory factor (IRF) activation, and to induce apoptosis. Ligand binding to these receptors results in TRIF recruitment through its TIR domain. Distinct protein-interaction motifs allow recruitment of the effector proteins TBK1, TRAF6 and RIPK1, which in turn, lead to the activation of transcription factors IRF3 and IRF7, NF-kappa-B and FADD respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIF
Applications Western Blot
Classification Monoclonal
Clone OTI1G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TICAM1
Gene Accession No. Q8IUC6
Gene Alias AW046014; AW547018; hCG_39321; HSD34; IIAE6; MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB; Putative NF-kappa-B-activating protein 502H; RNF36; TI; Ticam1; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; TIR domain-containing adaptor inducing interferon-beta; TIR-domain containing adaptor inducing interferon-beta; toll like receptor adaptor molecule 1; toll-interleukin 1 receptor domain-containing adapter protein inducing interferon-beta; toll-interleukin I receptor domain containing adaptor molecule 1; toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1; TRIF
Gene Symbols TICAM1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 120-420 of TICAM1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148022
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.