Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIF Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA595078

Catalog No. PIPA595078


Only null left
Add to Cart

Description

Description

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.

Involved in innate immunity against invading pathogens. Adapter used by TLR3 and TLR4 (through TICAM2) to mediate NF-kappa-B and interferon-regulatory factor (IRF) activation, and to induce apoptosis. Ligand binding to these receptors results in TRIF recruitment through its TIR domain. Distinct protein-interaction motifs allow recruitment of the effector proteins TBK1, TRAF6 and RIPK1, which in turn, lead to the activation of transcription factors IRF3 and IRF7, NF-kappa-B and FADD respectively.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIF
Polyclonal
Unconjugated
TICAM1
AW046014, AW547018, hCG_39321, HSD34, IIAE6, MyD88-3, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRB, Putative NF-kappa-B-activating protein 502H, RNF36, TI, Ticam1, TICAM-1, TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adapter protein inducing IFN-beta, TIR domain-containing adaptor inducing interferon-beta, TIR-domain containing adaptor inducing interferon-beta, toll like receptor adaptor molecule 1, toll-interleukin 1 receptor domain-containing adapter protein inducing interferon-beta, toll-interleukin I receptor domain containing adaptor molecule 1, toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta, toll-like receptor adaptor molecule 1, TRIF
Rabbit
Affinity chromatography
RUO
148022
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
500 μg/mL
PBS with 5MG BSA and 0.05MG sodium azide, 0.05MG thimerosal
Q8IUC6
TICAM1
A synthetic peptide corresponding to a sequence at the C-terminus of human TRIF (692-712aa NNHMWNQRGSQAPEDKTQEAE).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.