Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIF Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209515
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9515 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9515 Supplier Fabgennix Inc Supplier No. TRIF101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIF Polyclonal antibody specifically detects TRIF in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen TRIF
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene TICAM1
Gene Accession No. Q80UF7, Q8IUC6
Gene Alias AW046014; AW547018; hCG_39321; HSD34; IIAE6; MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB; Putative NF-kappa-B-activating protein 502H; RNF36; TI; Ticam1; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; TIR domain-containing adaptor inducing interferon-beta; TIR-domain containing adaptor inducing interferon-beta; toll like receptor adaptor molecule 1; toll-interleukin 1 receptor domain-containing adapter protein inducing interferon-beta; toll-interleukin I receptor domain containing adaptor molecule 1; toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1; TRIF
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 150-199 on human E3 ubiquitin-protein ligase RNF138 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106759, 148022, 363328
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.