Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIF (TICAM-1) Mouse anti-Human, PE, Clone: U19-709, BD

Catalog No. BDB566354 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566354 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566354 Supplier BD Biosciences Supplier No. 566354
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The U19-709 monoclonal antibody specifically recognizes TIR domain-containing adapter inducing IFN-β (TRIF),which is also known as TIR domain-containing adapter molecule 1 (TICAM-1). TRIF (TICAM-1) is an intracellular adaptor protein that contains a Toll/Interleukin-1 Receptor (TIR) homology domain. Double stranded RNA (dsRNA)-bound Toll-like receptor 3 (TLR3/CD283) or lipopolysaccharide (LPS)-triggered TLR4/CD284 can interact with TRIF (TICAM-1). This leads to downstream signaling cascades involving Interferon regulatory factors (eg, IRF3 or IRF7) or nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB) that induce Interferon-beta (IFN-β) expression.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIF (TICAM-1)
Classification Monoclonal
Clone U19-709
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TICAM1, PRVTIRB, MyD88-3, IIAE6
Host Species Mouse
Immunogen Human TRIF Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.