Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIF/TICAM1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB120277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120277 Supplier Novus Biologicals Supplier No. NB12013810AF647
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIF/TICAM1 Polyclonal specifically detects TRIF/TICAM1 in Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated.

Specifications

Antigen TRIF/TICAM1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse TRIF protein (between amino acids 100-180) [UniProt Q80UF7].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 148022
Target Species Mouse, Rat, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.