Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIF/TICAM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312427
This item is not returnable.
View return policy
Description
TRIF/TICAM1 Polyclonal antibody specifically detects TRIF/TICAM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
TRIF/TICAM1 | |
Polyclonal | |
Western Blot 1:500-1:750, ELISA 1:20000 | |
MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta | |
Synthetic peptide taken within amino acid region 150-199 on human E3 ubiquitin-protein ligase RNF138 protein. | |
100 μg | |
Signal Transduction | |
148022 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Unconjugated | |
Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction