Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018847 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
10 mL

Trifluoroacetic acid is a phytotoxic compound belonging to the class of perfluorinated acids (PFAs). It is an environmental contaminant, which contributes to the organo-fluorine burden in surface and ground waters.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 76-05-1
Vapor Pressure 97.5 mmHg (20°C)
Vapor Density 3.9 (vs. air)
Density 1.489 g/mL (at 20°C)
Physical Form Neat
Linear Formula CF3 COOH
MDL Number MFCD00004169
Melting Point -15.4°C
Boiling Point 72.4°C
Synonym TFA
Solubility Soluble in water
Type Analytical Standard
Quantity 10 mL
Formula Weight 114.02
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.