Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trifluoroacetic Acid, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle
Specifications
Formula Weight | 114.03 |
Grade | BAKER ANALYZED™ Reagent |
Quantity | 500 mL |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.53kg |
Chemical Name or Material | Trifluoroacetic Acid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction