Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, 99+%, for HPLC

Catalog No. p-4229347
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name 2,2,2-trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance 0.03 max. at 300 nm, 0.03 max. at 320 nm, 0.04 max. at 290 nm, 0.05 max. at 280 nm, 0.1 max. at 270nm, 1 max. at 260 nm
Melting Point -15°C
Boiling Point 72°C
Assay Percent Range 99+%
Linear Formula CF3CO2H
Beilstein 02,IV,458
Fieser 09,483; 17,340; 16,83; 15,338; 14,322; 12,529; 10,418; 07,388; 06,613; 05,695; 04,530; 03,305; 02,433; 01,1219; 08,503; 11,557
Merck Index 159847
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ether,acetone,ethanol,benzene,,ccl4,hexane,methanol,xylene and hexane
Formula Weight 114.02
Percent Purity 99+%
Grade HPLC
Density 1.5350 g/mL
Quantity 100 mL
Infrared Spectrum Conforms to Ref
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3
Residue after Evaporation 5 ppm max.
Specific Gravity 1.535
Vapor Pressure 107 mbar at 25°C
Chemical Name or Material Trifluoroacetic acid, For HPLC
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 200-929-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.