Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, HPLC Grade, 99.5+%

Catalog No. AA44630AE
Click to view available options
Quantity:
100 mL
500 mL
4 x 500 mL
This item is not returnable. View return policy

Catalog No. AA44630AE

Supplier: Thermo Scientific Chemicals 044630AE

This item is not returnable. View return policy

CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol

The combination of properties like solubility in most of the solvents, volatility, catalytic property and strong acidity with non-oxidizing nature makes it a widely used reagent in an organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and ceramic compounds. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. It finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name 2,2,2-trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15°C
Boiling Point 72°C to 73°C
Color Colorless
Assay Percent Range >99.5%
UN Number 2699
Beilstein 742035
Solubility Information Miscible with water,ether,acetone,ethanol,benzene,hexane and carbon tetrachloride.
Formula Weight 114.02
Grade HPLC
Density 1.48 g/cm3 at 20°C
Quantity 100 mL
Odor Pungent
Physical Form Liquid
Refractive Index 1.284
Sensitivity Hygroscopic
Vapor Pressure 8mm Hg at 20°C
Chemical Name or Material Trifluoroacetic Acid, Water 0.1%
Show More Show Less
Hazard Category H290-H314-H332-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: I
EINECSNumber 200-929-3
RTECSNumber AJ9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.