Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid lithium salt monohydrate, 95%, Thermo Scientific™

Catalog No. AC379770250
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C2 F3 Li O2 . H2 O
MDL Number MFCD00149663

Specifications

Color White
Assay Percent Range 0.95
Linear Formula CF3CO2Li·H2O
Formula Weight 137.97
Percent Purity ≥94%
Grade Pure
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Powder
Chemical Name or Material Trifluoroacetic acid lithium salt monohydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.