Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, MilliporeSigma™ Supelco™

Catalog No. 111022775
Click to view available options
Quantity:
10 mL
10 x 1 mL

Trifluoroacetic acid is commonly used as a mobile phase additive in liquid chromatography-mass spectrometry (LC-MS).

  • Extensive QC testing ensures the highest specifications
  • Optimized to improve ionization and resolution
  • Extremely low levels of inorganic and organic impurities
  • Manufactured specifically for accurate and fast LC-MS
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym TFA
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15.4°C (literature)
Boiling Point 72.4°C (literature)
Linear Formula CF3COOH
UN Number UN 2699 8/PGI
Formula Weight 114.02
Percent Purity ≥99% (GC)
Grade Eluent Additive for LC-MS
Density 1.489 g/mL (at 20°C (literature))
Quantity 10 mL
Refractive Index n20/D 1.3 (literature)
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Show More Show Less
Hazard Category H314 - H332 - H412
RTECSNumber AJ9625000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.