Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
GSA_VA

Catalog No. A11610X1AMP Shop All Fisherbrand Products
Click to view available options
Quantity:
0.5 mL
1 mL
2 mL
10 mL
50 mL
Packaging:
Amber Glass
Ampule
Ampules

Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL

Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL

  • Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
  • Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
  • Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
  • TFA is a common mobile phase additive that can function as an ion pair reagent

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance 0.003 AU max. at 254nm, 0.03 AU max. at 300nm, 0.06 AU max. at 230nm, 0.06 AU max. at 275nm, 0.18 AU max. at 220nm, 0.55 AU max. at 210nm
pH 0.45
Melting Point -15°C
Boiling Point 72°C
Color Colorless
Assay Percent Range ≥99.5 %
Linear Formula CF3COOH
Merck Index 15, 9847
Formula Weight 114.02
Percent Purity ≥99.5%
Grade OPTIMA™ LC-MS
Density 1.535g/cm³
Quantity 10 mL
Packaging Ampules
Physical Form Liquid
Residue after Evaporation 1ppm max.
Vapor Density 3.9
Vapor Pressure 107mbar 25
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe burns by all exposure routes. Inhalation may cause central nervous system effects. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.