Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, Silver Salt 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2966-50-9
C2AgF3O2
220.89
MFCD00013199
KZJPVUDYAMEDRM-UHFFFAOYSA-M
silver trifluoroacetate,silver 1+ trifluoroacetate,silver i 2,2,2-trifluoroacetate,silver mono trifluoroacetate,trifluoroacetic acid silver salt,silver 1+ ion trifluoroacetate,acetic acid, trifluoro-, silver 1+ salt,silver i trifluoroacetate,trifluoroacetic acid, silver salt,trifluoroacetic acid, silver 1+ salt
76299
silver;2,2,2-trifluoroacetate
C(=O)(C(F)(F)F)[O-].[Ag+]
2966-50-9
C2AgF3O2
220.89
MFCD00013199
KZJPVUDYAMEDRM-UHFFFAOYSA-M
silver trifluoroacetate,silver 1+ trifluoroacetate,silver i 2,2,2-trifluoroacetate,silver mono trifluoroacetate,trifluoroacetic acid silver salt,silver 1+ ion trifluoroacetate,acetic acid, trifluoro-, silver 1+ salt,silver i trifluoroacetate,trifluoroacetic acid, silver salt,trifluoroacetic acid, silver 1+ salt
76299
silver;2,2,2-trifluoroacetate
C(=O)(C(F)(F)F)[O-].[Ag+]
Melting Point 257.0°C to 260.0°C
Quantity 25g
Color Beige to Gray
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CF3CO2Ag
Fieser 01,1018; 07,323; 08,444; 11,471; 16,301; 17,154
Solubility Information Solubility in water: soluble
Formula Weight 220.89
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Trifluoroacetic acid, silver salt, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-004-0

TSCA : TSCA