Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trifluoromethanesulfonic Anhydride

Catalog No. 1761765G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-617-65G 5 g
17-617-650G 50 g
17-617-610G 10 g
17-617-61KG 1 kg
17-617-6250G 250 g
17-617-6500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Danger
Catalog No. 17-617-65G Supplier Sigma Aldrich Supplier No. 1761765G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trifluoromethanesulfonic anhydride can be used as: A trifluoromethylation reagent for the synthesis of trifluoromethylated compounds by direct introduction of CF3 group to (hetero)arenes. A reagent to prepare substituted tetrazoles from secondary amides using sodium azide. A reagent in Bischler-Napieraiski cyclization reaction along with 4-(N,N-dimethylamnino)pyridine.

Specifications

CAS 358-23-6
Density 1.677 g/mL at 25°C
Boiling Point 81°C to 83°C
Molecular Formula C2F6O5S2
Refractive Index n20/D 1.321
Linear Formula (CF3SO2)2O
MDL Number MFCD00000408
Quantity 5 g
Synonym Triflic anhydride
Molecular Weight (g/mol) 282.14 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.