Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trifluoromethanesulfonyl chloride, 98%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.
Solubility
Hydrolyzes with water.
Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.
Solubility
Hydrolyzes with water.
Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Density | 1.593 |
Boiling Point | 32°C |
Odor | Pungent |
Refractive Index | 1.334 |
Quantity | 100 g |
UN Number | UN3265 |
Beilstein | 1812016 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes with water. |
Formula Weight | 168.52 |
Show More |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 207-009-0 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction