Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trifluoromethoxy)benzene, 99%

Catalog No. p-7028374
Click to view available options
Quantity:
25 g
100 g

CAS: 456-55-3 | C7H5F3O | 162.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-55-3
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.11
MDL Number MFCD00040832
InChI Key GQHWSLKNULCZGI-UHFFFAOYSA-N
Synonym trifluoromethoxy benzene, 1-trifluoromethoxy benzene, benzene, trifluoromethoxy, phenyl trifluoromethyl ether, alpha,alpha,alpha-trifluoroanisole, trifluorophenoxymethane, pubchem7458, trifluoromethyloxybenzene, p-trifluoromethoxybenzene, trifluoromethoxy-benzene
PubChem CID 68010
IUPAC Name trifluoromethoxybenzene
SMILES FC(F)(F)OC1=CC=CC=C1

Specifications

Density 1.226
Boiling Point 102°C to 104°C
Flash Point 12°C (53°F)
Odor Aromatic
Refractive Index 1.406
Quantity 25 g
UN Number UN1993
Beilstein 2043132
Formula Weight 162.11
Percent Purity 99%
Chemical Name or Material (Trifluoromethoxy)benzene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-269-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.