Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, ACROS Organics™
Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
Specifications
Flash Point | -20°C |
Specific Gravity | 0.895 |
Boiling Point | 40.0°C |
Color | Colorless |
Density | 0.8950g/mL |
Packaging | Glass bottle |
Quantity | 5mL |
CAS Min % | 7.15,91.26 |
CAS Max % | 8.74,92.85 |
Linear Formula | (CH3)3SiCF3 |
Show More |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
Recommended Storage : Refrigerator +4°C