Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
81290-20-2,109-99-9
C4H9F3Si
142.2
MFCD00145454
MWKJTNBSKNUMFN-UHFFFAOYSA-N
trifluoromethyl trimethylsilane,trimethyl trifluoromethyl silane,trifluoromethyltrimethylsilane,ruppert's reagent,tms-cf3,tfmtms,silane, trimethyl trifluoromethyl,unii-a009786qpj,trimethylsilyl trifluoromethane,ruppert-prakash reagent
552549
trimethyl(trifluoromethyl)silane
C[Si](C)(C)C(F)(F)F
81290-20-2,109-99-9
C4H9F3Si
142.2
MFCD00145454
MWKJTNBSKNUMFN-UHFFFAOYSA-N
trifluoromethyl trimethylsilane,trimethyl trifluoromethyl silane,trifluoromethyltrimethylsilane,ruppert's reagent,tms-cf3,tfmtms,silane, trimethyl trifluoromethyl,unii-a009786qpj,trimethylsilyl trifluoromethane,ruppert-prakash reagent
552549
trimethyl(trifluoromethyl)silane
C[Si](C)(C)C(F)(F)F
Specific Gravity 0.895
Flash Point -20°C
Boiling Point 40.0°C
Density 0.8950g/mL
Quantity 100mL
Color Colorless
Packaging Glass bottle
CAS Min % 7.15,91.26
CAS Max % 8.74,92.85
Linear Formula (CH3)3SiCF3
Merck Index 15, 9851
Formula Weight 142.2
Physical Form Liquid
Chemical Name or Material (Trifluoromethyl)trimethylsilane, 0.5M solution in THF
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

Recommended Storage : Refrigerator +4°C