Learn More
Promega Corporation Triglyceride-Glo Assay
Catalog No. PRJ3160 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Detection Method:
Bioluminescence
Catalog No. Detection Method
PRJ3160 Bioluminescence
1 options
Catalog No. PRJ3160 Supplier Promega Corporation Supplier No. J3160
Detect Triglyceride from a variety of biological samples works with lysed cells, tissues, medium, and serum samples. No organic extraction, extreme heat, or centrifugation required, with linear triglyceride detection up to 80 μM.
  • Detects triglyceride levels from various biological samples.
  • No organic extraction, extreme heat, or centrifugation required.
  • Works with lysed cells, tissues, medium, and serum samples.
  • Linear triglyceride detection up to 80 μM.
  • Quick and simple assay protocol.
  • Produces reliable and reproducible results.
  • Suitable for research and diagnostic applications.
  • Cost-effective and efficient solution for triglyceride detection.
  • Ensures high sensitivity and specificity.
  • Compatible with various sample types.
Specifications
Detection Method Bioluminescence
Type Luminescent Detection Assay
Druggable Target Triglycerides