Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triiron dodecacarbonyl, 99%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
17685-52-8
C12Fe3O12
503.66
MFCD00135617
triiron dodecarbonyl
17685-52-8
C12Fe3O12
503.66
MFCD00135617
triiron dodecarbonyl
Specific Gravity 1.99
Stabilizer 5 to 10% MeOH
Color Black
Quantity 5g
Elemental Analysis 98.5% min.
Packaging Glass bottle
Density 1.9900g/mL
Infrared Spectrum Authentic
Melting Point 140.0°C
Assay Percent Range 99%
Linear Formula Fe3(CO)12
Fieser 04,534; 05,706; 07,144; 08,41
Solubility Information Solubility in water: insoluble
Formula Weight 503.66
Physical Form Powder or Crystals
Percent Purity 99%
Chemical Name or Material Triiron dodecacarbonyl
Show More Show Less

GHS H Statement
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and k

GHS Signal Word: Warning

EINECSNumber : 241-668-5