Learn More
Triisopropyl borate, 98+%
Description
Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.
Solubility
Miscible with ether, ethanol, isopropanol and benzene.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Decomposes in water.
Specifications
Specifications
| Melting Point | -59°C |
| Density | 0.815 |
| Boiling Point | 139°C to 141°C |
| Flash Point | 17°C (62°F) |
| Odor | Alcohol-like |
| Refractive Index | 1.376 |
| Linear Formula | [(CH3)2CHO]3B |
| Quantity | 500 mL |
| UN Number | UN2616 |
| Beilstein | 1701469 |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIISOPROPYL BORATE |
| EINECSNumber | 226-529-9 |
| RTECSNumber | ED5950000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.