Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triisopropyl borate, 98+%
Click to view available options
Quantity:
80 g
750 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -59°C |
Color | Colorless |
Density | 0.8140g/mL |
Boiling Point | 140°C |
Flash Point | 17°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.3755 to 1.3775 |
Linear Formula | [(CH3)2CHO]3B |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | GHS Signal Word: Danger |
missing translation for 'healthHazard2' | GHS H Statement: Highly flammable liquid and vapor. |
missing translation for 'healthHazard3' | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 226-529-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction