Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropyl borate, 98+%, Acros Organics

Click to view available options
Quantity:
80g
750g
Packaging:
Glass bottle
5419-55-6
C9H21BO3
188.07
MFCD00008872
NHDIQVFFNDKAQU-UHFFFAOYSA-N
triisopropyl borate,isopropyl borate,triisopropoxyborane,triisopropoxyboron,boron isopropoxide,triisopropyl orthoborate,boric acid triisopropyl ester,triisopropoxy borane,boron triisopropoxide,boric acid h3bo3 , tris 1-methylethyl ester
21531
tripropan-2-yl borate
B(OC(C)C)(OC(C)C)OC(C)C
5419-55-6
C9H21BO3
188.07
MFCD00008872
NHDIQVFFNDKAQU-UHFFFAOYSA-N
triisopropyl borate,isopropyl borate,triisopropoxyborane,triisopropoxyboron,boron isopropoxide,triisopropyl orthoborate,boric acid triisopropyl ester,triisopropoxy borane,boron triisopropoxide,boric acid h3bo3 , tris 1-methylethyl ester
21531
tripropan-2-yl borate
B(OC(C)C)(OC(C)C)OC(C)C
Flash Point 17°C
Boiling Point 140°C
Specific Gravity 0.814
Refractive Index 1.3755 to 1.3775
Melting Point -59°C
Density 0.8140g/mL
Infrared Spectrum Authentic
Quantity 750g
Color Colorless
Packaging Glass bottle
Assay Percent Range 98% min. (GC)
Linear Formula [(CH3)2CHO]3B
Beilstein 01, 363
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 188.07
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Triisopropyl borate
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 226-529-9