Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropylsilane, 98%

Catalog No. p-7045147
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 6485-79-6 | C9H22Si | 158.36 g/mol

Triisopropylsilane is used as a protecting group in peptide synthesis and is also used as a reducing agent for the selective reduction of anomeric C-phenyl ketals. It is used in the selective silylation of primary hydroxyl groups without affecting the secondary hydroxyl group. It is used in the preparation of anti-1,2-diols catalyzed by a Ni(O) N-heterocyclic carbine complex with high diastereoselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triisopropylsilane is used as a protecting group in peptide synthesis and is also used as a reducing agent for the selective reduction of anomeric C-phenyl ketals. It is used in the selective silylation of primary hydroxyl groups without affecting the secondary hydroxyl group. It is used in the preparation of anti-1,2-diols catalyzed by a Ni(O) N-heterocyclic carbine complex with high diastereoselectivity.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-79-6
Molecular Formula C9H22Si
Molecular Weight (g/mol) 158.36
MDL Number MFCD00009657
InChI Key YDJXDYKQMRNUSA-UHFFFAOYSA-N
Synonym triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
PubChem CID 6327611
SMILES CC(C)[SiH](C(C)C)C(C)C

Specifications

Boiling Point 166°C
Quantity 5 g
Linear Formula [(CH3)2CH]3SiH
UN Number UN1993
Beilstein 1733718
Flash Point 35°C (95°F)
Solubility Information Immiscible with water.
IUPAC Name tris(propan-2-yl)silane
Formula Weight 158.36
Percent Purity 98%
Odor Mild
Refractive Index 1.434
Density 0.773
Chemical Name or Material Triisopropylsilane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.