Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triisopropylsilane, 98%, ACROS Organics™
Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
Description
Reduction reagent
Specifications
Boiling Point | 84.0°C to 86.0°C (35.0mmHg) |
Specific Gravity | 0.773 |
Packaging | Glass bottle |
Flash Point | 37°C |
Refractive Index | 1.4340 to 1.4360 |
Density | 0.7730g/mL |
Quantity | 10g |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | [(CH3)2CH]3SiH |
Beilstein | 04,IV,3907 |
Show More |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning