Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM Antibody (488806), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4708V
Description
TRIM Monoclonal antibody specifically detects TRIM in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
TRIM | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
488806 | |
Western Blot, Intracellular Staining by Flow Cytometry | |
pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062 | |
E. coli-derived recombinant human TRIM, Thr50-Asn186, Accession # Q6PIZ9 | |
0.1 mL | |
Adaptive Immunity, Immunology | |
50852 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction