Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM Antibody (488806), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4708MFV450
Description
TRIM Monoclonal antibody specifically detects TRIM in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| TRIM | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 488806 | |
| Western Blot, Intracellular Staining by Flow Cytometry | |
| pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062 | |
| E. coli-derived recombinant human TRIM, Thr50-Asn186, Accession # Q6PIZ9 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 50852 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction