Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM Antibody (TRIM-04), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500485APC
Description
TRIM Monoclonal specifically detects TRIM in Human samples. It is validated for Flow Cytometry.Specifications
TRIM | |
Monoclonal | |
APC | |
pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The antibody TRIM-04 reacts with T cell receptor-interacting molecule (TRIM), a 30 kDa adaptor protein expressed by T cells. | |
Store at 4C in the dark. |
Flow Cytometry | |
TRIM-04 | |
Flow Cytometry | |
TRAT1 | |
Recombinant fragment, corresponding to amino acids 29-186 of Human TRIM. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
50852 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction