Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM Antibody (TRIM-04), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500485PEATT594
Description
TRIM Monoclonal specifically detects TRIM in Human samples. It is validated for Flow Cytometry.Specifications
TRIM | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
TRAT1 | |
Recombinant fragment, corresponding to amino acids 29-186 of Human TRIM. | |
0.1 mL | |
Primary | |
The antibody TRIM-04 reacts with T cell receptor-interacting molecule (TRIM), a 30 kDa adaptor protein expressed by T cells. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
TRIM-04 | |
Flow Cytometry | |
pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Immunology | |
50852.0 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction