Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM Mouse anti-Human, Mouse, Alexa Fluor™ 647, Clone: TRIM-4 (also known as TRIM-04), BD Pharmingen™

Catalog No. BDB568558 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568558 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB568558 Supplier BD Biosciences Supplier No. 568558
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRIM Monoclonal antibody specifically detects TRIM in Human, Mouse samples. It is validated for Intracellular staining (flow cytometry) (Routinely Tested)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM
Applications Flow Cytometry
Classification Monoclonal
Clone TRIM-4 (also known as TRIM-04)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias TRAT1; TRIM; T cell receptor interacting molecule; TCRIM
Host Species Mouse
Immunogen TRIM Cytoplasmic Sequence AA 29-186
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.