Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551706 Supplier Invitrogen™ Supplier No. PA551706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQDKMYSYSS DHTRVDEYYI EDTPIYGNLD DMISEPMDEN CYEQMKARPE KSVNKMQEAT PSAQATNETQ MCYASLDHSV KGKRRKPRKQ NTHFSDKDGD EQLHAI Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 58%.

TRIM (T cell receptor-interacting molecule), also known as TRAT1 (T cell receptor associated transmembrane adaptor 1) is a 30 kDa protein expressed by T cells as a cystein-linked homodimer. It associates with TCR-CD3-zeta-chain complex and becomes phosphorylated by Src-family kinases. TRIM is potentially involved in negative regulation of TCR-mediated signaling, but its role remains unclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRAT1
Gene Accession No. Q6PIZ9
Gene Alias C030046M14Rik; HSPC062; pp29/30; T cell receptor associated transmembrane adaptor 1; T cell receptor interacting molecule; T-cell receptor interacting molecule; T-cell receptor-associated transmembrane adapter 1; T-cell receptor-interacting molecule; TCRIM; Trat1; TRIM
Gene Symbols TRAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM. Recombinant protein control fragment (Product #RP-102133).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.