Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987238
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7238 Supplier Bioss Supplier No. BS6395R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by the RFP2 gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies near the nucleus, but its function is unknown. The gene is located on chromosome 13 within the minimal deletion region for B-cell chronic lymphocytic leukemia.

Specifications

Antigen TRIM13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TRIM13
Gene Accession No. O60858
Gene Alias 3110001L12Rik; B-cell chronic lymphocytic leukemia tumor suppressor Leu5; BOS_12360; CAR; CLL-associated RING finger; DLEU5; E3 ubiquitin-protein ligase TRIM13; LEU5; Leukemia-associated protein 5; Putative tumor suppressor RFP2; Ret finger protein 2; Rfp2; RING finger protein 77; RING-type E3 ubiquitin transferase TRIM13; RNF77; TRIM13; tripartite motif containing 13; tripartite motif protein 13; tripartite motif-containing 13; Tripartite motif-containing protein 13
Gene Symbols TRIM13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TRIM13/LEU5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10206
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.