Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531058 Supplier Invitrogen™ Supplier No. PA531058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: TRIM15-transfected 293T. Predicted reactivity: Pig (83%), Rhesus Monkey (97%), Chimpanzee (100%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene TRIM15
Gene Accession No. Q9C019
Gene Alias 1810012B10Rik; LOC100910806; RING finger protein 93; RNF93; TRIM15; tripartite motif containing 15; tripartite motif protein 15; tripartite motif-containing 15; tripartite motif-containing protein 15; tripartite motif-containing protein 15-like; zinc finger protein 178; Zinc finger protein B7; ZNF178; ZNFB7
Gene Symbols TRIM15
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TRIM15. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89870
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.