Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059772
Description
TRIM16 Polyclonal specifically detects TRIM16 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| TRIM16 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| DKFZp434O1826, EBBPtripartite motif-containing 16, Estrogen-responsive B box protein, tripartite motif containing 16, tripartite motif-containing protein 16 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10626 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2 - 5 ug/mg lysate | |
| NP_006461.3 | |
| TRIM16 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human tripartite motif-containing 16 (estrogen-responsive B box protein) using the numbering given in entry NP_006461.3 (GeneID 10626). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction