Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM16 Monoclonal Antibody (5F4)
GREENER_CHOICE

Catalog No. PIMA521106
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521106 Supplier Invitrogen™ Supplier No. MA521106
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LDAARRDKEA ELQCTQLDLE RKLKLNENAI SRLQANQKSV LVSVSEVKAV AEMQFGELLA AVRKAQANVM LFLEEKEQAA LSQANGIKAH LEYRSAEMEK SKQELERMA.

This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM16
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5F4
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Trim16
Gene Accession No. O95361
Gene Alias 9130006M08Rik; AI482483; E3 ubiquitin-protein ligase TRIM16; EBBP; Estrogen-responsive B box protein; TRIM16; tripartite motif containing 16; tripartite motif protein 16; tripartite motif-containing 16; tripartite motif-containing protein 16
Gene Symbols Trim16
Host Species Mouse
Immunogen TRIM16 (NP_006461, 165 a.a. ~ 273 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10626
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.